missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate Heptahydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | MgSO4 · 7H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 246.47g/mol |
Percent Purity | ≥98% |
Grade | ACS Reagent |
Vapor Pressure | 0.1mmHg (20°C) |
Density | Vapor Density: <0.01 (vs air) |
Chemical Name or Material | Magnesium Sulfate Heptahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'