missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Heptahydrate, ACS Reagent, Honeywell Fluka™

Catalog Number p-7071915
Click to view available options
Quantity:
25 g
500 g
1 kg
This item is not returnable. View return policy
10034-99-8
H14MgO11S
246.47
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
24843
CHEBI:31795
magnesium;sulfate;heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10034-99-8
H14MgO11S
246.47
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
24843
CHEBI:31795
magnesium;sulfate;heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 25 g
Linear Formula MgSO4 · 7H2O
UN Number NONH for all modes of transport
Formula Weight 246.47g/mol
Percent Purity ≥98%
Grade ACS Reagent
Vapor Pressure 0.1mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium Sulfate Heptahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'