Learn More
Magnesium silicide, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Magnesium silicide is used to produce alloys such as aluminum alloys. It is also used in food, beverages, as a flavor enhancer and as an intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1102°C |
Color | Blue |
Physical Form | Pieces |
Quantity | 10 g |
Assay Percent Range | 99.99% (Metals basis) |
Solubility Information | Insoluble in water and denser than water. |
Formula Weight | 76.71 |
Sensitivity | Air and moisture sensitive |
Density | 2 |
Chemical Name or Material | Magnesium silicide |