missing translation for 'onlineSavingsMsg'
Learn More

Magnesium silicide, Blue 3-12mm Pieces, 99.99% (Metals basis), Alfa Aesar™

Air and Moisture Sensitive; Decomposes in water, HCl

Brand:  Alfa Aesar 045518.18

Additional Details : CAS Number : 22831-39-6 Weight : 0.05000kg Transport : UN number : 2624 Chem class : 4.3 Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 15472327

  • 1163.44 DKK / 50g


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

22831-39-6
76.695
YTHCQFKNFVSQBC-UHFFFAOYSA-N
89858
Mg2Si
MFCD00016202
magnesium silicide, dimagnesium silicide, dimagnesium monosilicide, unii-475e6fmg3k, magnesiumsilicid, magnesium, compd. with silicon 2:1, magnesium silicide, powder, magnesium silicide-45 my, magnesium silicide un2624 dangerous when wet
[Mg]=[Si]=[Mg]

Specifications

Magnesium silicide
99.99% (Metals basis)
MFCD00016202
Insoluble in water and denser than water.
[Mg]=[Si]=[Mg]
89858
3-12mm Pieces
Air and moisture sensitive
1102°C
22831-39-6
Mg2Si
magnesium silicide, dimagnesium silicide, dimagnesium monosilicide, unii-475e6fmg3k, magnesiumsilicid, magnesium, compd. with silicon 2:1, magnesium silicide, powder, magnesium silicide-45 my, magnesium silicide un2624 dangerous when wet
YTHCQFKNFVSQBC-UHFFFAOYSA-N
76.695
76.71
2
Blue
50g
Videos
SDS Documents