missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Phosphate Dibasic Trihydrate, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
30 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | MgHPO4 · 3H2O |
UN Number | NONH for all modes of transport |
IUPAC Name | magnesium;hydrogen phosphate;trihydrate |
Formula Weight | 174.33g/mol |
Percent Purity | ≥98% |
Grade | puriss. special |
Density | 2.123g/mL at 25°C |
Chemical Name or Material | Magnesium Phosphate Dibasic Trihydrate, Meets analytical specification of DAB |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'