missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Phosphate Dibasic Trihydrate, Honeywell Fluka™

Catalog Number p-7071716
Click to view available options
Quantity:
1 kg
30 kg
This item is not returnable. View return policy
7782-75-4
H7MgO7P
174.33
MFCD00150390
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
16204530
O.O.O.[Mg++].OP([O-])([O-])=O
This item is not returnable. View return policy
7782-75-4
H7MgO7P
174.33
MFCD00150390
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
16204530
O.O.O.[Mg++].OP([O-])([O-])=O

Puriss special, meets analytical specification of DAB,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 1 kg
Linear Formula MgHPO4 · 3H2O
UN Number NONH for all modes of transport
IUPAC Name magnesium;hydrogen phosphate;trihydrate
Formula Weight 174.33g/mol
Percent Purity ≥98%
Grade puriss. special
Density 2.123g/mL at 25°C
Chemical Name or Material Magnesium Phosphate Dibasic Trihydrate, Meets analytical specification of DAB
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'