missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Oxide, Extra Pure, Heavy, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Specifications
Boiling Point | 3600°C |
Melting Point | 2800°C |
Color | White |
pH | 10.3 |
Physical Form | Powder/Solid |
Quantity | 250 g |
Formula Weight | 40.29g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 0 hPa at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'