Få mere at vide
Magnesium Monoperoxyphthalic Acid Hexahydrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Magnesium Monoperoxyphthalic Acid Hexahydrate |
| CAS | 84665-66-7 |
| Molekylær formel | C16H22MgO16 |
| Mængde | 100 g |
| Synonym | (T-4)-Bis[2-carboxybenzenecarboperoxoato(2-)-kOO,kO']magnesate(2-) Hydrogen Hydrate (1:2:6), 2-Carboxybenzenecarboperoxoic Acid Magnesium Salt (2:1) Hexahydrate, 2-Carboxybenzenecarboperoxoic Acid MagnesiumSalt Hydrate (2:1:6), Magnesium Bis(monoperoxyphthalate) Hexahydrate, MMPP, Magnesium Monoperoxyphthalate Hexahydrate |
| InChI formel | InChI=1S/2C8H6O5.Mg.6H2O/c2*9-7(10)5-3-1-2-4-6(5)8(11)13-12;;;;;;;/h2*1-4,12H,(H,9,10);;6*1H2/q;;+2;;;;;;/p-2 |
| SMIL | O.O.O.O.O.O.[Mg+2].OOC(=O)c1ccccc1C(=O)[O-].OOC(=O)c2ccccc2C(=O)[O-] |
| IUPAC navn | magnesium;2-carbonoperoxoylbenzoate;hexahydrate |
| Molekylvægt (g/mol) | 494.64 |
| Formel vægt | 494.0758 |