Learn More
Magnesium Chloride, Sigma-Aldrich™
Product Code. 15876530
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876530 500 g 500g
15866530 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15876530 Supplier Sigma Aldrich Supplier No. 8147330500
 Restricted Item
Forventet forsendelse: 18-11-2026
Anhydrous for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7786-30-3
Specifications
Boiling Point 1412°C/1013 hPa
Melting Point 712°C
Quantity 500 g
Density 2.32 g/cm3 at 20°C
MDL Number MFCD00011106
Synonym Magnesium chloride, Magnesium dichloride, Magnesium chloride
Molecular Weight (g/mol) 95.21
Grade Research
Chemical Name or Material Magnesium Chloride