missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Magnesium chloride hexahydrate, 99%, for biochemistry

Catalog Number p-4813527
Click to view available options
Quantity:
500 g
2.5 kg
7791-18-6
Cl2H12MgO6
203.30
MFCD00149781
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
7791-18-6
Cl2H12MgO6
203.30
MFCD00149781
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 117.0°C
Quantity 500 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula MgCl2·6H2O
Merck Index 15,5726
Solubility Information Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 203.31
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'