missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Acetate Tetrahydrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | (CH3COO)2Mg · 4H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3730605 |
Formula Weight | 214.45g/mol |
Percent Purity | ≥98% |
Grade | ACS Reagent |
Chemical Name or Material | Magnesium Acetate Tetrahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'