Få mere at vide
Magnesium 2-Aminoacetate, TRC
Artikelnummer. 30010266
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010266 50 mg
30003776 100 mg
30010170 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010266 Leverandør Toronto Research Chemicals Leverandørnr. TRCB47500350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14783-68-7
Molekylær formel C4H8MgN2O4
Molekylvægt (g/mol) 172.42
Synonym Magnesium Glycinate
IUPAC navn magnesium;2-aminoacetate
SMIL [Mg+2].NCC([O-])=O.NCC([O-])=O
Tekniske data
Kemisk navn eller materiale Magnesium 2-Aminoacetate
Mængde 50 mg
InChI formel InChI=1S/2C2H5NO2.Mg/c2*3-1-2(4)5;/h2*1,3H2,(H,4,5);/q;;+2/p-2
Formel vægt 172.0334