Få mere at vide
Tocris Bioscience™ Mag-Fura-2 AM
Beskrivelse
Mag-Fura-2 AM is a cell-permeable fluorescent magnesium ion (Mg2+) indicator. It shows selectivity for Mg2+ over other bivalent ions (Zn2+, Cu2+ and Ca2+). It exhibits >13,000-fold selectivity for Mg2+ ions over Ca2+ ions (Mg2+ Kd = 1.9 nM vs Ca2+ Kd = 25 μM). Excitation and emission maxima (λ) are 369 and 511 nm, respectively. Mag-Fura-2 AM can be used as an indirect indicator of ATP consumption/production.
It is recommended to prepare stock solutions in DMSO.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bis(acetoxymethyl) 2,2 '-((5-(2-(acetoxymethoxy)-2-oxoethoxy)-2-(5-((acetoxymethoxy)carbonyl)oxazol-2-yl)benzofuran-6-yl)azanediyl)diacetate |
| CAS | 130100-20-8 |
| Mængde | 1 mg |
| Mål | Ion Indicators |
| Molekylær formel | C30H30N2O19 |
| Renhed | 0.95 |