Learn More
Karl Hecht™ MACRO Stainless Steel Blood Sedimentation Apparatus
Product Code. 10185321
Change view
Click to view available options
Holds:
Pipettes (10)
Unit Size:
Each
Product Code. Holds Unit Size
10185321 Pipettes (10) Each
1 options
Product Code. 10185321 Supplier Karl Hecht™ Supplier No. 40367010
  • ONLINE EXCLUSIVE PRICE
    1621.46 DKK
    1258.37 DKK / Pakke med 1
    Spare 363.09 DKK
    22% Slukket
 Restricted Item
Forventet forsendelse: 22-01-2027
A complete unit with tilting device for rapid sedimentation.
  • With 10 fused graduation pipettes
  • Includes blood sedimentation pipettes no. 40380010
  • Protection caps for pipettes
  • Blood mixing tubes with rubber stoppers no. 40379010
  • Special rubber caps no. 40377015
  • Directions for use in an individual carton
Specifications
Holds Pipettes (10)
Material Stainless Steel
For Use With (Application) Rapid blood sedimentation
Quantity Each
Includes Pipettes, protecting caps, mixing tubes, rubber caps