Få mere at vide
Macitentan-d4, TRC
Artikelnummer. 30466851
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30466851 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466851 Leverandør Toronto Research Chemicals Leverandørnr. TRCM10500710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Macitentan-d4
CAS 1258428-05-5
Renhedsgrad noter HPLC
Molekylær formel C19H16D4Br2N6O4S
Mængde 10 mg
Synonym N-[5-(4-Bromophenyl)-6-[2-[(5-bromo-2-pyrimidinyl)oxy]ethoxy]-4-pyrimidinyl]-N'- propyl-Sulfamided4, ACT 064992-d4
InChI formel InChI=1S/C19H20Br2N6O4S/c1-2-7-26-32(28,29)27-17-16(13-3-5-14(20)6-4-13)18(25-12-24-17)30-8-9-31-19-22-10-15(21)11-23-19/h3-6,10-12,26H,2,7-9H2,1H3,(H,24,25,27)/i8D2,9D2
SMIL BrC1=CN=C(OC([2H])([2H])C([2H])([2H])OC2=C(C3=CC=C(Br)C=C3)C(NS(=O)(NCCC)=O)=NC=N2)N=C1
Molekylvægt (g/mol) 592.3
Procent renhed >95