missing translation for 'onlineSavingsMsg'
Learn More

M-Xylene, ReagentPlus™, 99%, Honeywell™

Catalog Number p-7120608
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
108-38-3
C8H10
106.17
MFCD00008536
IVSZLXZYQVIEFR-UHFFFAOYSA-N
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
7929
CHEBI:28488
1,3-xylene
CC1=CC(C)=CC=C1
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
108-38-3
C8H10
106.17
MFCD00008536
IVSZLXZYQVIEFR-UHFFFAOYSA-N
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
7929
CHEBI:28488
1,3-xylene
CC1=CC(C)=CC=C1

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -48°C
Color Colorless
Linear Formula C6H4(CH3)2
UN Number UN1307
Beilstein 605441
Formula Weight 106.17g/mol
Percent Purity 99%
Grade ReagentPlus™
Density 0.868g/mL at 25°C
Boiling Point 139°C
Quantity 1 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Ignition Point 982°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.497(lit.)
Vapor Pressure 16mmHg (37.7°C)
Chemical Name or Material M-Xylene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'