Få mere at vide
m-Xylene-D10, TRC
Artikelnummer. 30454177
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30454177 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454177 Leverandør Toronto Research Chemicals Leverandørnr. TRCX749417100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale m-Xylene D10
CAS 116601-58-2
Molekylær formel C8 2H10
Mængde 100 mg
Synonym m-Xylene D10, m-Xylene-d10 (6CI), 4,6-Di(methyl-d3)benzene-1,2,3,5-d4, 1,2,3,5-Tetradeuterio-4,6-bis(trideuteriomethyl)benzene, Perdeuterated 1,3-dimethylbenzene, Perdeuterated m-xylene, Benzene-1,2,3,5-d4','4,6-di(methyl-d3)-
InChI formel InChI=1S/C8H10/c1-7-4-3-5-8(2)6-7/h3-6H,1-2H3/i1D3,2D3,3D,4D,5D,6D
SMIL [2H]c1c([2H])c(c([2H])c(c1[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,2,3,5-tetradeuterio-4,6-bis(trideuteriomethyl)benzene
Molekylvægt (g/mol) 116.227
Formel vægt 116.141