Få mere at vide
m-Toluic-d7 Acid, CDN
Artikelnummer. 30313259
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30313259 1 g -
30324585 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313259 Leverandør CDN Leverandørnr. CDND63381G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale m-Toluic-d7 Acid
CAS 733046-94-1
Molekylær formel C8 2H7 H O2
Mængde 1 g
InChI formel InChI=1 S/C8H8O2/c1-6-3-2-4-7(5-6)8(9)10/h2-5 H,1H3,(H,9,10)/i1D3,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c(C(=O)O)c([2 H])c(c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,3,4,6-tetradeuterio-5-(trideuteriomethyl)benzoic acid
Molekylvægt (g/mol) 143.191
Formel vægt 143.096 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity