Få mere at vide
m-Terphenyl-d14, CDN
Artikelnummer. 30318673
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318673 0.5 g 0.5g
30312785 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318673 Leverandør CDN Leverandørnr. CDND54050.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale m-Terphenyl-D14
CAS 17714-84-0
Molekylær formel C18 D14
Mængde 0.5 g
Synonym m-Terphenyl-d14 (7 CI, 8 CI), 1,3-Terphenyl-d14, 3-(Phenyl-d5)-1,1'-biphenyl-2,2',3',4,4',5,5',6,6'-d9, 1,1':3',1''-Terphenyl-d14
InChI formel InChI=1 S/C18H14/c1-3-8-15(9-4-1)17-12-7-13-18(14-17)16-10-5-2-6-11-16/h1-14 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c([2 H])c([2 H])c([2 H])c(c2[2 H])c3c([2 H])c([2 H])c([2 H])c([2 H])c3[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-[2,3,4,6-tetradeuterio-5-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene
Molekylvægt (g/mol) 244.39
Formel vægt 244.1974 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity