Learn More
Brady™ M Series Indoor/Outdoor Industrial Vinyl Label Supply
This durable, low-shrink vinyl with ultra-aggressive adhesive is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
348.00 DKK
Specifications
Product Type | Label |
---|---|
Material | Vinyl |
Print Technology | Thermal Transfer |
Autoclavable | Not Autoclavable |
Description | Label |
Product Code | Brand | Dimensions (L x W) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Color | Price | Quantity & Availability | ||||
15143359
|
Brady™
131607 |
25 ft. x 1 in. | White on Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15173349
|
Brady™
131597 |
20 ft. x 1 in. | Black on Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces--including outdoors
- Conforms and adheres to difficult surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
- Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
- All-in-one cartridge includes ribbon and label supply
Specifications
Label | |
Thermal Transfer | |
Label | |
Cartridge, Ribbon |
Vinyl | |
Not Autoclavable | |
Gloss |