missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific™ M.R.S. Agar
GREENER_CHOICE

Artikelnummer. 15299822
Klik for at se tilgængelige muligheder
Mængde:
500 g
2.5 kg
Pakningsstørrelse:
2.5kg
500g
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 15299822

Brand: Thermo Scientific™ CM0361R

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Isolate and enumerate lactic acid bacteria from food samples with this medium.

Isolate and enumerate lactic acid bacteria from food with Thermo Scientific™ Oxoid™ M.R.S. Agar (Dehydrated). This medium is designed to support profuse growth of all strains of lactobacilli, even the difficult and slow-growing strains of Lactobacillus brevis and Lactobacillus fermenti.

M.R.S. Agar is designed to encourage the growth of the 'lactic acid bacteria' which includes species of the following genera: Lactobacillus, Streptococcus, Pediococcus, and Leuconostoc. All these species can produce lactic acid in considerable amounts.

  • Superior performance over the tomato juice medium of Briggs1 and the meat extract tomato juice medium of de Man
  • Allows growth of growth of all strains of lactobacilli, especially the difficult and slow-growing strains of Lactobacillus brevis and Lactobacillus fermenti

'Lactic acid bacteria' are Gram-positive, catalase and oxidase negative and are fastidious in their nutritional requirements. Growth is enhanced considerably by microaerobic conditions. Generally, the 'lactic acid bacteria' show delayed growth and smaller colony size than other microorganisms. They may be overgrown in non-selective media, especially if incubation is required for 2-4 days.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Tekniske data

Certificeringer/compliance Industrial Reference: AFNOR, IDF, NMKL standards.
Form Powder
Produkttype Agar
Mængde 2.5 kg
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.