Få mere at vide
m-Nitrobenzoic-d4 Acid, TRC
Artikelnummer. 30480319
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30480319 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480319 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49470210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale m-Nitrobenzoic-d4 Acid
CAS 78399-78-7
Molekylær formel C7 D4 H N O4
Mængde 10 mg
Synonym Benzoic-2,3,4,6-d4 acid','5-nitro- (9CI), 3-Nitro-2,4,5,6-tetradeuteriobenzoic acid, 5-Nitrobenzoic-2,3,4,6-d4 acid, 3-Nitrobenzoic-2,4,5,6-d4 acid, m-Nitrobenzoic acid-d4, 3-Nitrobenzoic Acid-D4, 3-Nitrobenzoic-d4 Acid, m-Nitrobenzoic-d4 Acid
InChI formel InChI=1S/C7H5NO4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H,(H,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C(=O)O)c([2H])c(c1[2H])[N+](=O)[O-]
IUPAC navn 2,3,4,6-tetradeuterio-5-nitrobenzoic acid
Molekylvægt (g/mol) 171.144
Formel vægt 171.047