Få mere at vide
m-Nifedipine, TRC
Artikelnummer. 30137320
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137320 1 g
17981795 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137320 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4570151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21881-77-6
Molekylær formel C17 H18 N2 O6
Molekylvægt (g/mol) 346.33
Synonym Dimethyl 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Nitrendipine Imp. B (EP), Nicardipine Hydrochloride Imp. C (EP), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 3,5-dimethyl ester, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, dimethyl ester (9CI), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(m-nitrophenyl)-, dimethyl ester (8CI), 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester, BAY-a 4339, Dimethyl 4-(m-nitrophenyl)-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, m-Nifedipine
IUPAC navn dimethyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMIL COC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC)C
Tekniske data
Kemisk navn eller materiale Dimethyl 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Mængde 1 g
InChI formel InChI=1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-6-5-7-12(8-11)19(22)23/h5-8,15,18H,1-4H3
Formel vægt 346.1165