Få mere at vide
m-Cresol-d8, CDN
Artikelnummer. 30324676
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324676 0.5 g
30319985 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324676 Leverandør CDN Leverandørnr. CDND35560.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metacresol-D8
CAS 302911-90-6
Molekylær formel C7 D8 O
Mængde 0.5 g
Synonym Phen-2,3,4,6-d4-ol-d, 5-(methyl-d3)- (9 CI), 3-(Methyl-d3)phenol-hydroxy,2,4,5,6-d5, m-Cresol-d7, 1-(Hydroxy-d)-3-(methyl-d3)benzene-2,4,5,6-d4, 3-Cresol-d8, m-Hydroxytoluene-d8, Metacresol-D8
InChI formel InChI=1 S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8 H,1H3/i1D3,2 D,3 D,4 D,5 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])c([2 H])c(c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,2,3,5-tetradeuterio-4-deuteriooxy-6-(trideuteriomethyl)benzene
Molekylvægt (g/mol) 116.1871
Formel vægt 116.1077 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity