Få mere at vide
m-Cresol-d7, TRC
Artikelnummer. 30365635
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365635 50 mg
30354708 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365635 Leverandør Toronto Research Chemicals Leverandørnr. TRCC78191750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202325-51-7
Molekylær formel C7 D7 H O
Molekylvægt (g/mol) 115.18
Synonym 3-Methylphenol-d7, 1-Hydroxy-3-methylbenzene-d7, 3-Hydroxytoluene-d7, 3-Methylphenol-d7, NSC 8768-d7, m-Cresylic Acid-d7, m-Hydroxytoluene-d7, m-Kresol-d7, m-Methylphenol-d7, m-Oxytoluene-d7, m-Toluol-d7, meta-Cresol-d7
IUPAC navn 2,3,4,6-tetradeuterio-5-(trideuteriomethyl)phenol
SMIL [2H]c1c([2H])c(O)c([2H])c(c1[2H])C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale m-Cresol-d7
Mængde 50 mg
InChI formel InChI=1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3/i1D3,2D,3D,4D,5D
Formel vægt 115.1015