missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ LZIP Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17110254 Shop alle Thermo Scientific produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17110254 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17110254 Leverandør Invitrogen™ Leverandørnr. PA5112390

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

LZIP Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds to the cAMP-responsive element, an octameric palindrome. The protein interacts with host cell factor C1, which also associates with the herpes simplex virus (HSV) protein VP16 that induces transcription of HSV immediate-early genes. This protein and VP16 both bind to the same site on host cell factor C1. It is thought that the interaction between this protein and host cell factor C1 plays a role in the establishment of latency during HSV infection. An additional transcript variant has been identified, but its biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen LZIP
Applikationer ELISA, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Koncentration 1.06 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CREB3
Genadgangsnr. O43889
Gene Alias AU044960; AW538053; basic leucine zipper protein; C80076; cAMP responsive element binding protein 3; cAMP-responsive element-binding protein 3; CREB3; CREB-3; cyclic AMP response element (CRE)-binding protein/activating transcription factor 1; cyclic AMP-responsive element-binding protein 3; leucin zipper proitein; Leucine zipper protein; Luman; LZIP; LZIP-1; LZIP-2; N-terminal Luman; Processed cyclic AMP-responsive element-binding protein 3; sLZIP; small leucine zipper protein; transcription factor LZIP; transcription factor LZIP-alpha
Gensymboler CREB3
Værtsarter Rabbit
Immunogen Recombinant Human Cyclic AMP-responsive element-binding protein 3 protein (1-230AA).
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 10488
Målarter Human
Indhold og opbevaring -20°C or -80°C if preferred
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.