missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1345.00 DKK - 3105.00 DKK
Tekniske data
| Antigen | Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B |
|---|---|
| Fortynding | Western Blot 1:500, ELISA 1:100-1:200 |
| Applikationer | Western Blot, ELISA |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18312167
|
Novus Biologicals
NBP3-18678-25UL |
25 μg |
1345.00 DKK
25µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18399324
|
Novus Biologicals
NBP3-18678-100UL |
100 μg |
3105.00 DKK
100µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B in Mouse samples. It is validated for Western Blot, ELISATekniske data
| Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Epigenetics | |
| Whole antiserum | |
| CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1 | |
| Polyclonal antibody raised in rabbit against mouse Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B (Jumonji, AT rich interactive domain 1B), using a KLH-conjugated synthetic peptides containing an amino acid sequence from the central part of the protein (1). | |
| Whole Antiserum |
| Western Blot 1:500, ELISA 1:100-1:200 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse | |
| Q9UGL1 | |
| 10765 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel