missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Antibody (PSH03-97), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32495
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Monoclonal antibody specifically detects Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B in Human samples. It is validated for Immunoprecipitation,Western Blot
Tekniske data
| Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunoprecipitation 1-2ug/sample | |
| CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1 | |
| Synthetic peptide within Human Lysine (K)-specific Demethylase 5B/Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B aa 960-1,009/ 1,544. (Uniprot: Q9UGL1) | |
| 100 μL | |
| Cancer, Epigenetics | |
| 10765 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| PSH03-97 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion