Få mere at vide
(+)-Lycopsamine N-Oxide (>85%), TRC
Artikelnummer. 30011813
Skift visning
Mængde:
0.5 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011813 5 mg
30005775 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011813 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4875305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 95462-15-0
Molekylær formel C15 H25 N O6
Molekylvægt (g/mol) 315.36
Synonym Butanoic acid, 2,3-dihydroxy-2-(1-methylethyl)-, (2,3,5,7a-tetrahydro-1-hydroxy-1H-pyrrolizin-7-yl)methyl ester, N-oxide, [1R-[1α,7(2S*,3S*),7aβ]]-, Butanoic acid, 2,3-dihydroxy-2-(1-methylethyl)-, [(1R,7aR)-2,3,5,7a-tetrahydro-1-hydroxy-4-oxido-1H-pyrrolizin-7-yl]methyl ester, (2S,3S)- (9CI), Lycopsamine N-oxide
IUPAC navn [(7R,8R)-7-hydroxy-4-oxido-5,6,7,8-tetrahydro-3H-pyrrolizin-4-ium-1-yl]methyl (2S)-2-hydroxy-2-[(1S)-1-hydroxyethyl]-3-methylbutanoate
SMIL CC(C)[C@](O)([C@H](C)O)C(=O)OCC1=CC[N+]2([O-])CC[C@@H](O)[C@@H]12
Tekniske data
Kemisk navn eller materiale Lycopsamine N-oxide
Mængde 5 mg
InChI formel InChI=1S/C15H25NO6/c1-9(2)15(20,10(3)17)14(19)22-8-11-4-6-16(21)7-5-12(18)13(11)16/h4,9-10,12-13,17-18,20H,5-8H2,1-3H3/t10-,12+,13+,15-,16?/m0/s1
Formel vægt 315.1682