missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Ly-6G/Ly-6C Antibody (RB6-8C5), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-00441UV
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Tekniske data
| Ly-6G/Ly-6C | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LY6G | |
| Normal mouse bone marrow. | |
| 0.1 mL | |
| Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell | |
| 546644 | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| RB6-8C5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| GR, Gr-1, Ly-6, Ly6G | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion