Få mere at vide
LY 368962, TRC
Artikelnummer. 30483380
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483380 100 mg
30474124 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30483380 Leverandør Toronto Research Chemicals Leverandørnr. TRCL486700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 193281-05-9
Molekylær formel C19 H21 N5 O7
Molekylvægt (g/mol) 431.4
Synonym L-Glutamic acid, N-[4-[3-(2,4-diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)-3-oxopropyl]benzoyl]-, (2S)-2-[[4-[3-(2,6-Diamino-4-oxo-1H-pyrimidin-5-yl)-3-oxo-propyl]benzoyl]amino]pentanedioic acid
IUPAC navn (2S)-2-[[4-[3-(2,6-diamino-4-oxo-1H-pyrimidin-5-yl)-3-oxopropyl]benzoyl]amino]pentanedioic acid
SMIL NC1=NC(=O)C(=C(N)N1)C(=O)CCc2ccc(cc2)C(=O)N[C@@H](CCC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale LY 368962
Mængde 100 mg
InChI formel InChI=1S/C19H21N5O7/c20-15-14(17(29)24-19(21)23-15)12(25)7-3-9-1-4-10(5-2-9)16(28)22-11(18(30)31)6-8-13(26)27/h1-2,4-5,11H,3,6-8H2,(H,22,28)(H,26,27)(H,30,31)(H5,20,21,23,24,29)/t11-/m0/s1
Formel vægt 431.1441