missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
LY-364947, MedChemExpress
Description
LY-364947 (HTS466284) is a potent ATP-competitive inhibitor of TGFβR-I with IC50 of 59 nM, and exhibits 7-fold selectivity over TGFβR-II.
Specifications
Specifications
| Kemisk navn eller materiale | LY-364947 |
| Fysisk form | Solid |
| CAS | 396129-53-6 |
| Farve | Light Yellow |
| Mængde | 50 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C17H12N4 |
| Til brug med (applikation) | Cancer-Kinase/protease |
| Synonym | HTS466284 |
| Opløselighedsinformation | DMSO : 25 mg/mL (91.81 mM; Need ultrasonic and warming) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?