Få mere at vide
Tocris Bioscience™ LY 2955303
Beskrivelse
LY 2955303 is a high affinity and selective RARγ antagonist (Ki values are 1.1 nM, >1.7 μM and >2.9 μM for RARγ, RARα and RARβ respectively). Displays analgesic efficacy in a mouse model of osteoarthritis-like joint pain.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[5-[3,5-Bis(1,1-dimethylethyl)phenyl]-1-[4-[(4-methyl-1-piperazinyl)carbonyl]phenyl]-1H-pyrazol-3-yl]benzoic acid |
| CAS | 1433497-19-8 |
| Mængde | 5 mg |
| Opløselighed | Soluble to 20 mM in DMSO |
| Molekylær formel | C36H42N4O3 |
| Formel vægt | 578.74 |
| Produkttype | Antagonist |
| Renhed | 98% |