Få mere at vide
Tocris Bioscience™ LY 266097 hydrochloride
GREENER_CHOICE
Artikelnummer. 18065222
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18065222 50 mg
18034682 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18065222 Leverandør Tocris Bioscience™ Leverandørnr. 4081/50
Selective 5-HT2B receptor antagonist
Selective 5-HT2B receptor antagonist (pKi = 9.3). More than 100-fold selective over 5-HT2A and 5-HT2C. Attenuates amphetamine-induced locomotion in the rat.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 172895-39-5
Molekylær formel C21H24Cl2N2O2
Tekniske data
Kemisk navn eller materiale LY 266097 hydrochloride
Mængde 50 mg
Formel vægt 407.33
Renhed >99%
Molekylvægt (g/mol) 407.335