missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 266097 hydrochloride
GREENER_CHOICE

Product Code. 18034682
Change view
Click to view available options
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18034682 10 mg
18065222 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18034682 Supplier Tocris Bioscience™ Supplier No. 4081/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective 5-HT2B receptor antagonist

Selective 5-HT2B receptor antagonist (pKi = 9.3). More than 100-fold selective over 5-HT2A and 5-HT2C. Attenuates amphetamine-induced locomotion in the rat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172895-39-5
Molekylær formel C21H24Cl2N2O2

Specifications

Kemisk navn eller materiale LY 266097 hydrochloride
Mængde 10 mg
Formel vægt 407.33
Renhed >99%
Molekylvægt (g/mol) 407.335
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.