Få mere at vide
Welch™ LVS 110 Z Laboratory Vacuum System
Shop alle Welch Ilmvac produkter
Dybde (metrisk):
310 mm
Pakningsstørrelse:
Hver
Beskrivelse
- Available with 2 or 3 pumping stages to generate vacuum as low as 2 mbar (1.5 torr)
- Flow rate choices from 20 to 238L/min.
- Modular design with options to complement your process
- Digital control options and on board accessories tailor the LVS to your needs
- Resistant to acid and solvent vapors
- Wetted parts in PTFE, PVDF, PEEK, PP and glass
- Ergonomic design places controls and features where they are needed
- Frees up hood or bench space
- Less maintenance compared to other technologies saves time and money
- Dry vacuum allows cleaner workplace and environment
- Uncontaminated pumping of gases results in cleaner samples and no risk of oil back-streaming
LVS systems are available with a range of vacuum control options; unregulated, manually regulated and three different digital control packages are available.
Tekniske data
Tekniske data
| Elektriske krav | 230 V 50/60 Hz |
| Dybde (metrisk) | 310 mm |
| Model- | LVS 110 Z |
| Flowhastighed | 1.0/1.1 m³/hr. |
| Frekvens | 50/60 Hz |
| Samlet højde (metrisk) | 445 mm |
| Samlet bredde (metrisk) | 360 mm |
| Pumpetype | Laboratory Vacuum System |
| Maksimalt tryk | <8 mbar |
| Spænding | 230 V |