Learn More
Lutetium Solution for ICP, 100mg/L, HNO3 Matrix, Fisher Chemical™
Catalog Number 12917283
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 100mg/L
Quantity 100 mL
Solution Contains HNO3