missing translation for 'onlineSavingsMsg'
Få mere at vide

(+)-Lupanine Perchlorate, TRC

Artikelnummer. 30012795
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
2.5 mg
10 mg
Förpackningsstorlek
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Mængde unitSize
30012795 1 mg -
30007268 10 mg -
30012005 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30012795 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCL4748301MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7400-11-5
Molekylær formel C15 H24 N2 O . Cl H O4
Molekylvægt (g/mol) 348.82
Synonym 7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, dodecahydro-, (7S,7aR,14S,14aS)-, perchlorate (1:1), 7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, dodecahydro-, (7S,7aR,14S,14aS)-, monoperchlorate (9CI), 7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, dodecahydro-, [7S-(7α,7aα,14α,14aβ)]-, monoperchlorate, Lupanine, monoperchlorate (8CI), Lupanine perchlorate, (7S,7aR,14S,14aS)-Dodecahydro-7,14-methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one perchlorate, (+)-2-Oxosparteine perchlorate, (+)-Lupanine perchlorate, 2-Oxosparteine perchlorate, D-(+)-Lupanine perchlorate, D-Lupanine perchlorate, Lupanin perchlorate, d-Lupanine perchlorate
SMIL OCl(=O)(=O)=O.O=C1CCC[C@@H]2[C@H]3C[C@@H](CN12)[C@@H]4CCCCN4C3

Specifikationer

Kemisk navn eller materiale D-(+)-Lupanine Perchlorate
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C15H24N2O.ClHO4/c18-15-6-3-5-14-11-8-12(10-17(14)15)13-4-1-2-7-16(13)9-11;2-1(3,4)5/h11-14H,1-10H2;(H,2,3,4,5)/t11-,12-,13-,14+;/m0./s1
Formel vægt 348.1452
Procent renhed >95%
Produkttitel
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.