Få mere at vide
(+)-Lupanine Perchlorate, TRC
Artikelnummer. 30007268
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30007268 10 mg -
30012005 2.5 mg 2.5mg
30012795 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007268 Leverandør Toronto Research Chemicals Leverandørnr. TRCL47483010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7400-11-5
Molekylær formel C15 H24 N2 O . Cl H O4
Molekylvægt (g/mol) 348.82
Synonym 7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, dodecahydro-, (7S,7aR,14S,14aS)-, perchlorate (1:1), 7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, dodecahydro-, (7S,7aR,14S,14aS)-, monoperchlorate (9CI), 7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, dodecahydro-, [7S-(7α,7aα,14α,14aβ)]-, monoperchlorate, Lupanine, monoperchlorate (8CI), Lupanine perchlorate, (7S,7aR,14S,14aS)-Dodecahydro-7,14-methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one perchlorate, (+)-2-Oxosparteine perchlorate, (+)-Lupanine perchlorate, 2-Oxosparteine perchlorate, D-(+)-Lupanine perchlorate, D-Lupanine perchlorate, Lupanin perchlorate, d-Lupanine perchlorate
SMIL OCl(=O)(=O)=O.O=C1CCC[C@@H]2[C@H]3C[C@@H](CN12)[C@@H]4CCCCN4C3
Tekniske data
Kemisk navn eller materiale D-(+)-Lupanine Perchlorate
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C15H24N2O.ClHO4/c18-15-6-3-5-14-11-8-12(10-17(14)15)13-4-1-2-7-16(13)9-11;2-1(3,4)5/h11-14H,1-10H2;(H,2,3,4,5)/t11-,12-,13-,14+;/m0./s1
Formel vægt 348.1452
Procent renhed >95%