missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Lung Specific Antigen Antibody (LSA120), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32870-0.1mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Lung Specific Antigen Monoclonal antibody specifically detects Lung Specific Antigen in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Tekniske data
| Lung Specific Antigen | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1 μg/mL, Immunohistochemistry-Frozen 0.5-1 μg/mL | |
| Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like | |
| Protein extract of human lung cells | |
| 0.1 mg | |
| Primary | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
| LSA120 | |
| Unconjugated | |
| PBS with 0.05% BSA. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion