Få mere at vide
Lumefantrine-d18, TRC
Artikelnummer. 30467971
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30467971 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467971 Leverandør Toronto Research Chemicals Leverandørnr. TRCL47400210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lumefantrine-d18
CAS 1185240-53-2
Molekylær formel C30H14D18Cl3NO
Mængde 10 mg
Synonym (9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-α-[(dibutylamino-d18)methyl]-9H-fluorene-4-methanol, 2-(Dibutylamino-d18)-1-[2,7-dichloro-9-[1-(4-chlorophenyl)meth-(Z)-
InChI formel InChI=1S/C30H32Cl3NO/c1-3-5-13-34(14-6-4-2)19-29(35)28-18-23(33)17-27-25(15-20-7-9-21(31)10-8-20)26-16-22(32)11-12-24(26)30(27)28/h7-12,15-18,29,35H,3-6,13-14,19H2,1-2H3/b25-15-/i1D3,2D3,3D2,4D2,5D2,6D2,13D2,14D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])N(CC(O)c1cc(Cl)cc2\C(=C/c3ccc(Cl)cc3)\c4cc(Cl)ccc4c12)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
Molekylvægt (g/mol) 547.05