Få mere at vide
Luliconazole, MedChemExpress
Artikelnummer. 16436436
Encompass_Preferred
Skift visning
Mængde:
50 mg
100 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16436436 100 mg
16432167 50 mg
16411126 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16436436 Leverandør Medchem Express Leverandørnr. HY14283
MedChemExpress Luliconazole (NND 502) is a topical antifungal imidazole antibiotic with broad-spectrum and potent antifungal activity. Luliconazole can be used for the research of skin infection, including dermatophytosis, tinea corporis, tinea pedis et al.
Luliconazole (NND 502) is a topical antifungal imidazole antibiotic with broad-spectrum and potent antifungal activity. Luliconazole can be used for the research of skin infection, including dermatophytosis, tinea corporis, tinea pedis et al.
Tekniske data
Kemisk navn eller materiale Luliconazole
Fysisk form Solid
CAS 187164-19-8
Farve Yellow
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C14H9Cl2N3S2
Til brug med (applikation) COVID-19-immunoregulation
Synonym NND 502
Opløselighedsinformation DMSO : 50 mg/mL (141.13 mM; Need ultrasonic)
SMIL ClC1=CC(Cl)=CC=C1[C@@H]2CS/C(S2)=C(C#N)\N3C=CN=C3
Molekylvægt (g/mol) 354.28
Formel vægt 354.28
Procent renhed 98.78%