Få mere at vide
MedChemExpress Lucigenin
Tekniske data
Tekniske data
| Indhold og opbevaring | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Produkttype | Dye Reagents |
| Mængde | 100 mg |
| Forsendelsestilstand | Room temperature |
| Opløselighed | DMSO : 62.5 mg/mL (122.43 mM; Need ultrasonic) |
| Mål | HBV |
| Molekylvægt (g/mol) | 510.5 g/mol |
| CAS | 2315-97-1 |
| Synonym | NSC-151912 ∣ L-6868 |