Få mere at vide
Lucanthone Hydrochloride, TRC
Artikelnummer. 30465770
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30465770 100 mg
1 options
Artikelnummer. 30465770 Leverandør Toronto Research Chemicals Leverandørnr. TRCL473700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lucanthone Hydrochloride
CAS 548-57-2
Molekylær formel C20 H24 N2 O S . Cl H
Mængde 100 mg
Synonym 9H-Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, monohydrochloride (9CI), Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, monohydrochloride (6CI,8CI), 1-[(2-Diethylaminoethyl)amino]-4-methylthioxanthen-9-one hydrochloride, 3735 R. P., 79T61, BW 57-233, Lucanthone hydrochloride, Lucanthone monohydrochloride, Miracil D, Miracol, Ms. 752, NSC 14574, Nilodin, RP 3735, Scapuren, Tixantone, 9H-Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, hydrochloride (1:1), Lucanthone Hydrochloride
InChI formel InChI=1S/C20H24N2OS.ClH/c1-4-22(5-2)13-12-21-16-11-10-14(3)20-18(16)19(23)15-8-6-7-9-17(15)24-20;/h6-11,21H,4-5,12-13H2,1-3H3;1H
SMIL Cl.CCN(CC)CCNc1ccc(C)c2Sc3ccccc3C(=O)c12
IUPAC navn 1-[2-(diethylamino)ethylamino]-4-methylthioxanthen-9-one;hydrochloride
Molekylvægt (g/mol) 376.94
Formel vægt 376.1376