Få mere at vide
Lucanthone Hydrochloride, Mikromol™
Beskrivelse
Lucanthone Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Lucanthone Hydrochloride |
| CAS | 548-57-2 |
| Analyt- eller komponentnavne | Lucanthone Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H24 N2 O S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 9H-Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, monohydrochloride (9CI), Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, monohydrochloride (6CI,8CI), 1-[(2-Diethylaminoethyl)amino]-4-methylthioxanthen-9-one hydrochloride, 3735 R. P., 79T61, BW 57-233, Lucanthone hydrochloride, Lucanthone monohydrochloride, Miracil D, Miracol, Ms. 752, NSC 14574, Nilodin, RP 3735, Scapuren, Tixantone, 9H-Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, hydrochloride (1:1), Lucanthone Hydrochloride |
| InChI formel | InChI=1S/C20H24N2OS.ClH/c1-4-22(5-2)13-12-21-16-11-10-14(3)20-18(16)19(23)15-8-6-7-9-17(15)24-20;/h6-11,21H,4-5,12-13H2,1-3H3;1H |
| SMIL | Cl.CCN(CC)CCNc1ccc(C)c2Sc3ccccc3C(=O)c12 |
| IUPAC navn | 1-[2-(diethylamino)ethylamino]-4-methylthioxanthen-9-one;hydrochloride |
| Vis mere |