missing translation for 'onlineSavingsMsg'
Learn More

Lubiprostone-d7, TRC

Product Code. 30469117
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30469117 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30469117 Supplier Toronto Research Chemicals Supplier No. TRCL4735035MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Lubiprostone-d7
CAS 1217675-13-2
Molekylær formel C20H25D7F2O5
Mængde 5 mg
Synonym (11α)-16,16-Difluoro-11-hydroxy-9,15-dioxo-prostan-1-oic Acid-d7, Ru 0211-d7
InChI formel InChI=1S/C20H32F2O5/c1-2-3-12-20(21,22)18(25)11-10-15-14(16(23)13-17(15)24)8-6-4-5-7-9-19(26)27/h14-15,17,24H,2-13H2,1H3,(H,26,27)/t14-,15-,17-/m1/s1/i1D3,2D2,3D2
SMIL O=C(C(F)(F)CC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])CC[C@@H]1[C@@H](CCCCCCC(O)=O)C(C[C@H]1O)=O
Molekylvægt (g/mol) 397.51
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.