missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ LTBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599878
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Antibody detects endogenous levels of total LTBP1.
The protein encoded by this gene belongs to the family of latent TGF-beta binding proteins. The secretion and activation of TGF-betas is regulated by their association with latency-associated proteins and with latent TGF-beta binding proteins. The product of this gene targets latent complexes of transforming growth factor beta to the extracellular matrix, where the latent cytokine is subsequently activated by several different mechanisms. Alternatively spliced transcript variants encoding different isoforms have been identified.
Tekniske data
| LTBP1 | |
| Polyclonal | |
| Unconjugated | |
| LTBP1 | |
| 9430031G15Rik; 9830146M04; b2b1000Clo; LanC (bacterial lantibiotic synthetase component C)-like 1; latent TGF beta binding protein; latent transforming growth factor beta binding protein 1; latent transforming growth factor binding protein 1; latent transforming growth factor binding protein-1; latent-transforming growth factor beta-binding protein 1; LTBP1; LTBP-1; MGC163161; Tgfb; TGF-beta-1-BP-1; TGF-beta1-BP-1; transforming growth factor beta-1-binding protein 1; Transforming growth factor beta-1-masking protein large subunit; transforming growth factor-beta (TGF-beta) masking protein large subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 268977, 4052, 59107 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q00918, Q14766, Q8CG19 | |
| LTBP1 | |
| A synthesized peptide derived from human LTBP1(Accession Q14766), corresponding to amino acid residues S1674-E1721. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion