missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ LSN 3318839

Artikelnummer. 30226366
Skift visning
Klik for at se tilgængelige muligheder
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
30226366 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30226366 Leverandør Tocris Bioscience™ Leverandørnr. 7959/5

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Glucagon-like peptide-1 receptor (GLP1-R) positive allosteric modulator

LSN 3318839 is a potent glucagon-like peptide-1 receptor (GLP1-R) positive allosteric modulator (EC50 = 14 nM). Acts as a molecular glue to stabilize peptide ligand and GLP1 receptor interaction. Potentiates activity of GLP-1 (7-36) (Cat. No. 2082) and the low-affinity metabolite GLP-1 (9-36) (Cat. No. 3266). LSN 3318839 alone or in combination with Sitagliptin (Cat. No. 6816) lowers glucose level in animal models of diabetes. LSN 3318839 is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifica

Kemisk navn eller materiale (R)-2-(2-(1-((R)-1-(2,6-Dichloro-3-cyclopropylphenyl)ethyl)-1H-imidazo[4,5-c]pyridin-6-yl)phenyl)propanoic acid
CAS 2764704-18-7
Mængde 5 mg
Mål Glucagon-Like Peptide 1 Receptor Modulators
Molekylær formel C26H23Cl2N3O2
Renhed 0.98
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.