missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC74A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-83163-0.1ML
Additional Details : Weight : 0.00970kg
Description
LRRC74A Polyclonal specifically detects LRRC74A in Human samples. It is validated for Western Blot.Specifications
LRRC74A | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
145497 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
chromosome 14 open reading frame 166B, hypothetical protein LOC145497 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human LRRC74A | |
0.1 mL | |
Primary | |
Human | |
Purified |