Få mere at vide
LRRC41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Artikelnummer. 16830005 Shop alle Proteintech produkter
Skift visning
Mængde:
20 μL
150 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16830005 20 μL
16820005 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16830005 Leverandør Proteintech Leverandørnr. 204571AP20UL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3, 000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis.
Tekniske data
Antigen LRRC41
Applikationer Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Koncentration 0.13 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LRRC41
Genadgangsnr. Q15345, Q5M9H1, Q8K1C9
Gene Alias LRRC41, MUF1, PP7759, Protein Muf1, RP4 636H5.2
Gensymboler Lrrc41
Værtsarter Rabbit
Immunogen LRRC41 Fusion Protein Ag14266
Oprensningsmetode Antigen Affinity Chromatography
Mængde 20 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 10489, 230654, 362566
Målarter Human, Rat, Mouse
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG